Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123060562-123060791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA8
Entrez Gene ID
3312
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
UniGene
Hs.180414
Gene Chromosome Location
Chr.11: 123057492-123062366 on Build GRCh38
Transcripts
NM_006597.5 NM_153201.3 XM_011542798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
924153 c.382A>G p.K128E Chr.11: 123060622-123060622 on Build GRCh38
924154 c.277A>G p.M93V Chr.11: 123060727-123060727 on Build GRCh38
5414060 c.378G>A p.K126K Chr.11: 123060626-123060626 on Build GRCh38
4018790 c.242C>G p.A81G Chr.11: 123060762-123060762 on Build GRCh38
4835734 c.219G>A p.L73L Chr.11: 123060785-123060785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201223120 Chr.11: 123060707-123060707 on Build GRCh38
rs140536048 Chr.11: 123060671-123060671 on Build GRCh38
rs11551608 Chr.11: 123060658-123060658 on Build GRCh38
rs746344123 Chr.11: 123060731-123060731 on Build GRCh38
rs755808417 Chr.11: 123060710-123060710 on Build GRCh38
rs768398499 Chr.11: 123060602-123060602 on Build GRCh38
rs751477695 Chr.11: 123060695-123060695 on Build GRCh38
rs200601870 Chr.11: 123060688-123060688 on Build GRCh38
rs759998595 Chr.11: 123060656-123060656 on Build GRCh38
rs748008431 Chr.11: 123060684-123060684 on Build GRCh38
rs781044959 Chr.11: 123060678-123060678 on Build GRCh38
rs780961303 Chr.11: 123060698-123060698 on Build GRCh38
rs777228127 Chr.11: 123060717-123060717 on Build GRCh38
rs374961418 Chr.11: 123060626-123060626 on Build GRCh38
rs1273833 Chr.11: 123060592-123060592 on Build GRCh38
rs772769841 Chr.11: 123060620-123060620 on Build GRCh38
rs147610984 Chr.11: 123060629-123060629 on Build GRCh38
rs774683371 Chr.11: 123060634-123060634 on Build GRCh38
rs747769427 Chr.11: 123060610-123060610 on Build GRCh38
rs371835507 Chr.11: 123060683-123060683 on Build GRCh38
rs11551601 Chr.11: 123060716-123060716 on Build GRCh38
rs766369333 Chr.11: 123060693-123060693 on Build GRCh38
rs775043697 Chr.11: 123060728-123060728 on Build GRCh38
rs769361497 Chr.11: 123060611-123060611 on Build GRCh38
rs780873363 Chr.11: 123060609-123060609 on Build GRCh38
rs760971468 Chr.11: 123060674-123060674 on Build GRCh38
rs879251486 Chr.11: 123060659-123060659 on Build GRCh38
rs570819499 Chr.11: 123060752-123060752 on Build GRCh38
rs764136878 Chr.11: 123060677-123060677 on Build GRCh38
rs748864676 Chr.11: 123060719-123060719 on Build GRCh38
rs879004891 Chr.11: 123060637-123060637 on Build GRCh38
rs770300995 Chr.11: 123060722-123060722 on Build GRCh38

View Full Product Details