Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 31247744-31248017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GLCT
Entrez Gene ID
145173
Gene Name
beta 3-glucosyltransferase
Gene Aliases
B3GALTL, B3GTL, B3Glc-T, Gal-T, beta3Glc-T
UniGene
Hs.13205
Gene Chromosome Location
Chr.13: 31199975-31332276 on Build GRCh38
Transcripts
NM_194318.3 XM_006719768.3 XM_011534936.1 XM_011534938.2 XM_017020395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279097 c.367A>G p.R123G Chr.13: 31247874-31247874 on Build GRCh38
5527221 c.374C>T p.S125L Chr.13: 31247881-31247881 on Build GRCh38
6609254 c.374C>A p.S125* Chr.13: 31247881-31247881 on Build GRCh38
6314138 c.348-5G>T p.? Chr.13: 31247850-31247850 on Build GRCh38
3700403 c.407G>T p.R136I Chr.13: 31247914-31247914 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs139934988 Chr.13: 31247891-31247891 on Build GRCh38
rs1036695673 Chr.13: 31247960-31247960 on Build GRCh38
rs775418673 Chr.13: 31247962-31247962 on Build GRCh38
rs761621191 Chr.13: 31247980-31247980 on Build GRCh38
rs141743580 Chr.13: 31247957-31247957 on Build GRCh38
rs917658694 Chr.13: 31247943-31247943 on Build GRCh38
rs371293220 Chr.13: 31247824-31247824 on Build GRCh38
rs1006887899 Chr.13: 31247790-31247790 on Build GRCh38
rs766485660 Chr.13: 31247901-31247903 on Build GRCh38
rs374438614 Chr.13: 31247804-31247804 on Build GRCh38
rs759575352 Chr.13: 31247945-31247945 on Build GRCh38
rs376926710 Chr.13: 31247898-31247898 on Build GRCh38
rs774100166 Chr.13: 31247977-31247977 on Build GRCh38
rs968996840 Chr.13: 31247838-31247838 on Build GRCh38
rs369251610 Chr.13: 31247928-31247928 on Build GRCh38
rs755455921 Chr.13: 31247906-31247906 on Build GRCh38
rs4943266 Chr.13: 31247855-31247855 on Build GRCh38
rs756854037 Chr.13: 31247871-31247871 on Build GRCh38
rs748444091 Chr.13: 31247920-31247920 on Build GRCh38
rs990546165 Chr.13: 31247914-31247914 on Build GRCh38
rs566881850 Chr.13: 31247794-31247794 on Build GRCh38
rs546325942 Chr.13: 31247885-31247885 on Build GRCh38
rs762077884 Chr.13: 31247978-31247978 on Build GRCh38
rs373567541 Chr.13: 31247947-31247947 on Build GRCh38
rs1020981140 Chr.13: 31247827-31247827 on Build GRCh38
rs376755630 Chr.13: 31247973-31247973 on Build GRCh38
rs751269401 Chr.13: 31247860-31247860 on Build GRCh38
rs768816639 Chr.13: 31247849-31247849 on Build GRCh38
rs767109506 Chr.13: 31247984-31247984 on Build GRCh38
rs779419580 Chr.13: 31247909-31247909 on Build GRCh38
rs202155585 Chr.13: 31247991-31247991 on Build GRCh38
rs185063097 Chr.13: 31247942-31247942 on Build GRCh38
rs754338962 Chr.13: 31247899-31247899 on Build GRCh38
rs776694081 Chr.13: 31247854-31247855 on Build GRCh38
rs750025878 Chr.13: 31247987-31247987 on Build GRCh38
rs548683413 Chr.13: 31247779-31247779 on Build GRCh38
rs765978204 Chr.13: 31247988-31247988 on Build GRCh38
rs755612098 Chr.13: 31247893-31247893 on Build GRCh38
rs775363299 Chr.13: 31247821-31247821 on Build GRCh38
rs1021979833 Chr.13: 31247803-31247803 on Build GRCh38
rs767361165 Chr.13: 31247967-31247967 on Build GRCh38

View Full Product Details