Gene Transcripts

Gene Symbol
Trim50
Entrez Gene ID
215061
Gene Name
tripartite motif-containing 50
Gene Aliases
Chromosome Location
Chr. 5: 135351872 - 135367903 on Build GRCm38
UniGene ID
Mm.261834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178240.2 NP_839971.1 6 1303
GenBank mRNA AY081947.1 - 6 1303
BC130022.1 - 6 1309 40140742

View Full Product Details