Gene Transcripts

Gene Symbol
Zfp58
Entrez Gene ID
238693
Gene Name
zinc finger protein 58
Gene Aliases
A530094I17Rik, Mfg-1, Mfg1, Rslcan5, Zfp817
Chromosome Location
Chr. 13: 67490167 - 67500522 on Build GRCm38
UniGene ID
Mm.478914
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007575.2 NP_001007576.1 4 1272
GenBank mRNA AK041250.1 - 4 1208
AK167399.1 - 4 1214

View Full Product Details