Gene Transcripts

Gene Symbol
Poc1b
Entrez Gene ID
382406
Gene Name
POC1 centriolar protein homolog B (Chlamydomonas)
Gene Aliases
4933430F16Rik, Wdr51b
Chromosome Location
Chr. 10: 99107171 - 99198074 on Build GRCm38
UniGene ID
Mm.440656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027740.6 NP_082016.1 8, 9 916
GenBank mRNA AK016991.1 - 7, 8 840
AK030180.1 - 7, 8 810
AK045302.1 - 8, 9 895
AK050233.1 - 8, 9 985
AK090075.1 - 8, 9 919
BC050888.1 - 7, 8 914
BC099926.1 - 8, 9 1003
BC144819.1 - 8, 9 1005
BC145801.1 - 8, 9 1005 40131104

View Full Product Details