Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028006.2 | NP_082282.1 | 11 | 1175 | |
GenBank mRNA | AK010005.1 | - | 11 | 1169 | |
AK040492.1 | - | 3 | 2733 | ||
AK041416.1 | - | 11 | 1141 | ||
AK041986.1 | - | 581 | |||
AK087403.1 | - | 10 | 954 | ||
AK148039.1 | - | 11 | 1175 | ||
BC038654.1 | - | 8 | 1140 | ||
BC062179.1 | - | 11 | 1172 | 6333817 |