Gene Transcripts

Gene Symbol
Olfr2
Entrez Gene ID
18317
Gene Name
olfactory receptor 2
Gene Aliases
I7, MOR103-15, Olfr41
Chromosome Location
Chr. 7: 107000875 - 107005558 on Build GRCm38
UniGene ID
Mm.246525
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010983.2 NP_035113.1 2 291
GenBank mRNA BC128014.1 - 1 29

View Full Product Details