Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025476.6 | NP_079752.3 | 4, 5 | 632 | |
GenBank mRNA | AK002442.1 | - | 4, 5 | 515 | |
AK009059.1 | - | 4, 5 | 490 | ||
AK010421.2 | - | 4, 5 | 509 | ||
AK029446.1 | - | 4, 5 | 492 | ||
AK160266.1 | - | 4, 5 | 513 | ||
BC025158.1 | - | 4, 5 | 505 | 4023319 | |
BC096046.1 | - | 4, 5 | 609 | 1397855 |