Gene Transcripts

Gene Symbol
Ofcc1
Entrez Gene ID
218165
Gene Name
orofacial cleft 1 candidate 1
Gene Aliases
opo
Chromosome Location
Chr. 13: 40001834 - 40361259 on Build GRCm38
UniGene ID
Mm.364096
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172143.2 NP_742155.2 2 86
GenBank mRNA AF548110.1 - 3 370
AF548111.1 - 2 106
EU683438.1 - 2 86
EU683439.1 - 2 86

View Full Product Details