Gene Transcripts

Gene Symbol
Gzmn
Entrez Gene ID
245839
Gene Name
granzyme N
Gene Aliases
GrN
Chromosome Location
Chr. 14: 56165786 - 56174612 on Build GRCm38
UniGene ID
Mm.207063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB049454.2 - 5 574
AB100426.1 - 5 574
BC120809.1 - 4 495 40129732

View Full Product Details