Gene Transcripts

Gene Symbol
Urm1
Entrez Gene ID
68205
Gene Name
ubiquitin related modifier 1 homolog (S. cerevisiae)
Gene Aliases
2900073H19Rik
Chromosome Location
Chr. 2: 29827389 - 29844996 on Build GRCm38
UniGene ID
Mm.20273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026615.4 NP_080891.1 3 183
GenBank mRNA AK012124.1 - 3 189
AK019352.1 - 3 189
AK049227.1 - 3 184
AK049849.1 - 3 184
AK210360.1 - 3 167
BC026994.1 - 3 175 5368627

View Full Product Details