Gene Transcripts

Gene Symbol
Olr49
Entrez Gene ID
293211
Gene Name
olfactory receptor 49
Gene Aliases
ratchr1-160568743-160569669_ORF
Chromosome Location
Chr. 1: 168028367 - 168029293 on Build Rnor_6.0
UniGene ID
Rn.141925
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000132.1 NP_001000132.1 1 505

View Full Product Details