Gene Transcripts

Gene Symbol
Pcdhgb8
Entrez Gene ID
364845
Gene Name
protocadherin gamma subfamily B, 8
Gene Aliases
Pcdhgb5
Chromosome Location
Chr. 18: 30909318 - 30971113 on Build Rnor_6.0
UniGene ID
Rn.8628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037159.1 NP_001032236.1 1 1113
GenBank mRNA AY574027.1 - 1 1113

View Full Product Details