Gene Transcripts

Gene Symbol
Prp2l1
Entrez Gene ID
287750
Gene Name
proline rich protein 2-like 1
Gene Aliases
PRP, PRP-2, Prp-5, Prp2l2
Chromosome Location
Chr. 10: 91610662 - 91690864 on Build Rnor_6.0
UniGene ID
Rn.77316
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768296.1 XP_008766518.1 3 11522
GenBank mRNA M86514.1 - 14897

View Full Product Details