Gene Transcripts
- Gene Symbol
- TMEM11
- Entrez Gene ID
- 8834
- Gene Name
- transmembrane protein 11
- Gene Aliases
- C17orf35, PM1, PMI
- Chromosome Location
- Chr. 17: 21197950 - 21214595 on Build GRCh38
- UniGene ID
- Hs.592945
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024547.1 |
|
2 |
650 |
|
GenBank mRNA |
DC364753.1 |
- |
2 |
216 |
|
- Gene Symbol
- ENO1
- Entrez Gene ID
- 2023
- Gene Name
- enolase 1
- Gene Aliases
- ENO1L1, HEL-S-17, MPB1, NNE, PPH
- Chromosome Location
- Chr. 1: 8861000 - 8879092 on Build GRCh38
- UniGene ID
- Hs.517145
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK298600.1 |
- |
|
216 |
|
View Full Product Details