Gene Transcripts
- Gene Symbol
- PWRN1
- Entrez Gene ID
- 791114
- Gene Name
- Prader-Willi region non-protein coding RNA 1
- Gene Aliases
- NCRNA00198
- Chromosome Location
- Chr. 15: 24558157 - 24587780 on Build GRCh38
- UniGene ID
- Hs.720900
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026646.1 |
|
3, 4 |
589 |
|
GenBank mRNA |
BC034815.1 |
- |
3, 4 |
589 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG717318.1 |
- |
3, 4 |
594 |
|
Chr. 15 - 24558157 - 24574647 [+] |
View Full Product Details