Gene Transcripts
- Gene Symbol
- MSX2P1
- Entrez Gene ID
- 55545
- Gene Name
- msh homeobox 2 pseudogene 1
- Gene Aliases
- HPX5, HSHPX5, MSX2P
- Chromosome Location
- Chr. 17: 58156959 - 58159120 on Build GRCh38
- UniGene ID
- Hs.381329
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002307.1 |
|
1 |
751 |
|
- Gene Symbol
- MSX2
- Entrez Gene ID
- 4488
- Gene Name
- msh homeobox 2
- Gene Aliases
- CRS2, FPP, HOX8, MSH, PFM, PFM1
- Chromosome Location
- Chr. 5: 174724495 - 174730899 on Build GRCh38
- UniGene ID
- Hs.89404
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X69295.1 |
- |
|
728 |
|
View Full Product Details