Gene Transcripts

Gene Symbol
NBPF4
Entrez Gene ID
148545
Gene Name
neuroblastoma breakpoint family member 4
Gene Aliases
Chromosome Location
Chr. 1: 108222464 - 108244081 on Build GRCh38
UniGene ID
Hs.511849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011540763.1 XP_011539065.1 16 3075
GenBank mRNA AK057395.1 - 14 2445

Gene Symbol
NBPF22P
Entrez Gene ID
285622
Gene Name
neuroblastoma breakpoint family member 22, pseudogene
Gene Aliases
Chromosome Location
Chr. 5: 86282444 - 86297547 on Build GRCh38
UniGene ID
Hs.449272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003719.2 8 2306
GenBank mRNA BC050328.1 - 8 2262

View Full Product Details