Gene Transcripts
- Gene Symbol
- TTTY7
- Entrez Gene ID
- 246122
- Gene Name
- testis-specific transcript, Y-linked 7 (non-protein coding)
- Gene Aliases
- CLONE795723, LINC00129, NCRNA00129, TTTY7A, TTTY7B, TTY7
- Chromosome Location
- Chr. Y: 6449468 - 6457906 on Build GRCh38
- UniGene ID
- Hs.407277
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF278762.1 |
- |
2 |
288 |
|
View Full Product Details