Gene Transcripts
- Gene Symbol
- Igkv4-86
- Entrez Gene ID
- 243451
- Gene Name
- immunoglobulin kappa variable 4-86
- Gene Aliases
- Gm459, kb4
- Chromosome Location
- Chr. 6: 68910411 - 68910938 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ372749.1 |
- |
|
191 |
|
DQ372755.1 |
- |
|
191 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AJ697689.1 |
- |
|
140 |
|
Chr. - - [-] |
GenBankmRNA |
AK157689.1 |
- |
|
915 |
|
Chr. - - [-] |
View Full Product Details