Gene Transcripts
- Gene Symbol
- RREB1
- Entrez Gene ID
- 6239
- Gene Name
- ras responsive element binding protein 1
- Gene Aliases
- FINB, HNT, LZ321, RREB-1, Zep-1
- Chromosome Location
- Chr. 6: 7107597 - 7251980 on Build GRCh38
- UniGene ID
- Hs.298248
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022160.1 |
- |
2 |
1459 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000451355.2 |
- |
2 |
1460 |
|
Chr. 6 - 7183083 - 7185287 [+] |
View Full Product Details