Gene Transcripts
- Gene Symbol
- TCIRG1
- Entrez Gene ID
- 10312
- Gene Name
- T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
- Gene Aliases
- ATP6N1C, ATP6V0A3, Atp6i, OC-116kDa, OC116, OPTB1, Stv1, TIRC7, Vph1, a3
- Chromosome Location
- Chr. 11: 68038995 - 68053839 on Build GRCh38
- UniGene ID
- Hs.495985
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000534517.1 |
- |
2 |
588 |
|
Chr. 11 - 68052823 - 68053762 [+] |
View Full Product Details