Gene Transcripts

Gene Symbol
PWRN1
Entrez Gene ID
791114
Gene Name
Prader-Willi region non-protein coding RNA 1
Gene Aliases
NCRNA00198
Chromosome Location
Chr. 15: 24558157 - 24587780 on Build GRCh38
UniGene ID
Hs.720900
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026646.1 2 244
GenBank mRNA BC034815.1 - 2 244


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY041248.1 - 2 206 Chr. 15 - 24558195 - 24574574 [+]
GENCODE ENST00000565241.1 - 2 184 Chr. 15 - 24558217 - 24568121 [+]
GenBankmRNA BG717318.1 - 2 249 Chr. 15 - 24558157 - 24574647 [+]
GENCODE ENST00000568045.5 - 3 464 Chr. 15 - 24533695 - 24587777 [+]
GenBankmRNA DB459584.1 - 2 184 Chr. 15 - 24558217 - 24568121 [+]
GENCODE ENST00000568609.5 - 2 249 Chr. 15 - 24558152 - 24587779 [+]
GenBankmRNA HY040849.1 - 2 176 Chr. 15 - 24558226 - 24568101 [+]

View Full Product Details