Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172514.3 | NP_766102.1 | 7 | 1069 | |
GenBank mRNA | AK088711.1 | - | 7 | 1070 | |
AK170268.1 | - | 7 | 1066 | ||
BC117823.1 | - | 7 | 1002 | 40092863 | |
BC117824.1 | - | 7 | 1002 | 40092864 |