Gene Transcripts

Gene Symbol
Stox1
Entrez Gene ID
216021
Gene Name
storkhead box 1
Gene Aliases
4732470K04Rik, Gm63
Chromosome Location
Chr. 10: 62659043 - 62726099 on Build GRCm38
UniGene ID
Mm.39821
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033260.1 NP_001028432.1 3 2388
GenBank mRNA AK028920.1 - 3 2155
AK133671.1 - 1 976
BC137978.1 - 3 2388 8860997

View Full Product Details