Gene Transcripts

Gene Symbol
PON3
Entrez Gene ID
5446
Gene Name
paraoxonase 3
Gene Aliases
Chromosome Location
Chr. 7: 95359872 - 95396375 on Build GRCh38
UniGene ID
Hs.440967
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000940.2 NP_000931.1 7 781
GenBank mRNA AF320003.1 - 7 756
AF329433.1 - 7 756
AK096114.1 - 2 2112
BC070374.1 - 8 767 4710338
CD014095.1 - 8 913
CD014097.1 - 7 769
CD014100.1 - 7 640
L48516.1 - 7 717

View Full Product Details