Gene Transcripts

Gene Symbol
Vmn2r11
Entrez Gene ID
384219
Gene Name
vomeronasal 2, receptor 11
Gene Aliases
EG384219
Chromosome Location
Chr. 5: 109046873 - 109059452 on Build GRCm38
UniGene ID
Mm.469885
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104622.1 NP_001098092.1 2 353

View Full Product Details