Gene Transcripts

Gene Symbol
Vmn2r90
Entrez Gene ID
626942
Gene Name
vomeronasal 2, receptor 90
Gene Aliases
EG626942
Chromosome Location
Chr. 17: 17669137 - 17734167 on Build GRCm38
UniGene ID
Mm.404019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104539.1 NP_001098009.1 3 1259

View Full Product Details