Gene Transcripts

Gene Symbol
SPP2
Entrez Gene ID
6694
Gene Name
secreted phosphoprotein 2
Gene Aliases
SPP-24, SPP24
Chromosome Location
Chr. 2: 234050671 - 234077134 on Build GRCh38
UniGene ID
Hs.444488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006944.2 NP_008875.1 2 251
XM_005246102.3 XP_005246159.1 2 282
XM_011511698.2 XP_011510000.1 2 282
XM_011511699.2 XP_011510001.1 2 282
XM_011511700.2 XP_011510002.1 2 282
GenBank mRNA AJ308099.1 - 2 251
BC069401.1 - 2 166 7262158
BC106705.1 - 2 166
BC112438.1 - 2 166
U20530.1 - 2 166

View Full Product Details