Gene Transcripts

Gene Symbol
Slco6b1
Entrez Gene ID
170925
Gene Name
solute carrier organic anion transporter family, member 6b1
Gene Aliases
GST-1, Tst1
Chromosome Location
Chr. 9: 104388232 - 104468001 on Build Rnor_6.0
UniGene ID
Rn.27679
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133412.1 NP_596903.1 10, 11 2007
XM_006245576.2 XP_006245638.1 10, 11 2077
GenBank mRNA AF321415.1 - 10, 11 2007

View Full Product Details