Gene Transcripts

Gene Symbol
Wnt5b
Entrez Gene ID
282582
Gene Name
wingless-type MMTV integration site family, member 5B
Gene Aliases
Chromosome Location
Chr. 4: 151500962 - 151516894 on Build Rnor_6.0
UniGene ID
Rn.58724
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100489.1 NP_001093959.1 4 738
XM_006237179.1 XP_006237241.1 3 616
GenBank mRNA AF481944.1 - 2 263
BC161860.1 - 881 7096336

View Full Product Details