Gene Transcripts

Gene Symbol
TNNC2
Entrez Gene ID
7125
Gene Name
troponin C2, fast skeletal type
Gene Aliases
Chromosome Location
Chr. 20: 45823214 - 45833329 on Build GRCh38
UniGene ID
Hs.182421
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003279.2 NP_003270.1 4 322
XM_011529031.2 XP_011527333.1 4 332
GenBank mRNA AK291323.1 - 4 320
AK300665.1 - 320
BC005323.1 - 4 320 3934188
BU854061.1 - 4 353
CR541864.1 - 3 256
CR541884.1 - 3 256
M22307.1 - 4 303
X07898.1 - 4 320

View Full Product Details