Gene Transcripts

Gene Symbol
tGap1
Entrez Gene ID
294892
Gene Name
GTPase activating protein testicular GAP1
Gene Aliases
Chromosome Location
Chr. 2: 92552805 - 92625921 on Build Rnor_6.0
UniGene ID
Rn.162562
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007635.2 NP_001007636.2 10 1054
XM_006232134.2 XP_006232196.1 12 1396
GenBank mRNA AY631396.1 - 10 1054
AY631398.1 - 3 198
AY631399.1 - 10 819
AY631400.1 - 10 1021

Gene Symbol
LOC100910852
Entrez Gene ID
100910852
Gene Name
uncharacterized LOC100910852
Gene Aliases
Chromosome Location
Chr. 2: 90211214 - 90251905 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760856.1 XP_008759078.1 1472

Gene Symbol
LOC102555809
Entrez Gene ID
102555809
Gene Name
rho GTPase-activating protein 20-like
Gene Aliases
Chromosome Location
Chr. 8: 17668536 - 17702279 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766004.1 XP_008764226.1 10 1698

View Full Product Details