Gene Transcripts

Gene Symbol
Tnfrsf12a
Entrez Gene ID
302965
Gene Name
tumor necrosis factor receptor superfamily, member 12a
Gene Aliases
Fn14
Chromosome Location
Chr. 10: 12995904 - 12997930 on Build Rnor_6.0
UniGene ID
Rn.105040
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181086.3 NP_851600.1 4 634
GenBank mRNA BC060537.1 - 4 638
FQ229590.1 - 4 634

View Full Product Details