Gene Transcripts

Gene Symbol
Hddc3
Entrez Gene ID
308758
Gene Name
HD domain containing 3
Gene Aliases
RGD1311839
Chromosome Location
Chr. 1: 142136429 - 142138737 on Build Rnor_6.0
UniGene ID
Rn.17508
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107528.1 NP_001100998.1 4 514
XM_006229431.1 XP_006229493.1 4 640

View Full Product Details