Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001108536.1 | NP_001102006.1 | 3, 4 | 421 | |
XM_006256528.2 | XP_006256590.1 | 4, 5 | 339 | ||
XM_006256529.1 | XP_006256591.1 | 3, 4 | 209 | ||
GenBank mRNA | BC158720.1 | - | 4, 5 | 323 |