Gene Transcripts

Gene Symbol
Sla2
Entrez Gene ID
499931
Gene Name
Src-like-adaptor 2
Gene Aliases
RGD1562071
Chromosome Location
Chr. 3: 152943953 - 153004259 on Build Rnor_6.0
UniGene ID
Rn.145609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172118.1 NP_001165589.1 5 446
XM_008762374.1 XP_008760596.1 7 2123

View Full Product Details