Gene Transcripts

Gene Symbol
Bsx
Entrez Gene ID
500982
Gene Name
brain specific homeobox
Gene Aliases
RGD1565120
Chromosome Location
Chr. 8: 45061670 - 45065430 on Build Rnor_6.0
UniGene ID
Rn.228217
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191995.1 NP_001178924.1 2, 3 513

View Full Product Details