Gene Transcripts
- Gene Symbol
- Creld1
- Entrez Gene ID
- 171508
- Gene Name
- cysteine-rich with EGF-like domains 1
- Gene Aliases
- AI843811
- Chromosome Location
- Chr. 6: 113483297 - 113493338 on Build GRCm38
- UniGene ID
- Mm.41593
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133930.1 |
NP_598691.1
|
8, 9 |
1244 |
|
GenBank mRNA |
AB009020.1 |
- |
8, 9 |
1249 |
|
AK050160.1 |
- |
1, 2 |
25 |
|
AK050455.1 |
- |
1, 2 |
25 |
|
AK077726.1 |
- |
8, 9 |
1389 |
|
AK181275.1 |
- |
3, 4 |
254 |
|
BC010804.1 |
- |
8, 9 |
1244 |
4239786 |
BC023893.1 |
- |
8, 9 |
1255 |
5310111 |
BC025932.1 |
- |
8, 9 |
1150 |
5102571 |
BC029065.1 |
- |
8, 9 |
1280 |
4935296 |
View Full Product Details