Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032108.3 | NP_115484.2 | 8 | 923 | |
XM_011527639.2 | XP_011525941.1 | 8 | 972 | ||
XM_011527640.2 | XP_011525942.1 | 8 | 715 | ||
XM_011527641.1 | XP_011525943.1 | 8 | 972 | ||
GenBank mRNA | AB022433.1 | - | 7 | 699 | |
AB209278.1 | - | 7 | 685 | ||
AF216389.1 | - | 7 | 664 | ||
AF293363.1 | - | 7 | 664 | ||
AK300037.1 | - | 8 | 923 | ||
AY358939.1 | - | 7 | 679 | ||
BC142617.1 | - | 8 | 866 | 40147342 | |
FR839673.1 | - | 7 | 661 |