Gene Transcripts

Gene Symbol
PDCD11
Entrez Gene ID
22984
Gene Name
programmed cell death 11
Gene Aliases
ALG-4, ALG4, NFBP, RRP5
Chromosome Location
Chr. 10: 103396615 - 103446295 on Build GRCh38
UniGene ID
Hs.239499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014976.1 NP_055791.1 5, 6 654
XM_005269647.3 XP_005269704.1 5, 6 694
XM_011539538.2 XP_011537840.1 5, 6 642
XM_011539539.2 XP_011537841.1 5, 6 642
XM_011539540.1 XP_011537842.1 4, 5 508
GenBank mRNA AK294971.1 - 5, 6 683
AK307418.1 - 5, 6 667
BC049838.1 - 5, 6 655
BC064486.1 - 5, 6 647
BC080560.1 - 5, 6 647
BC111040.1 - 5, 6 609
D80007.1 - 5, 6 607

View Full Product Details