Gene Transcripts
- Gene Symbol
- TEN1-CDK3
- Entrez Gene ID
- 100529145
- Gene Name
- TEN1-CDK3 readthrough (NMD candidate)
- Gene Aliases
- C17orf106-CDK3
- Chromosome Location
- Chr. 17: 75979217 - 76005999 on Build GRCh38
- UniGene ID
- Hs.706766
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037709.1 |
|
3 |
414 |
|
GenBank mRNA |
AK097104.1 |
- |
3 |
371 |
|
BX647274.1 |
- |
3 |
400 |
|
- Gene Symbol
- TEN1
- Entrez Gene ID
- 100134934
- Gene Name
- TEN1 CST complex subunit
- Gene Aliases
- C17orf106
- Chromosome Location
- Chr. 17: 75979217 - 76000586 on Build GRCh38
- UniGene ID
- Hs.593566
- Species
- Homo sapiens
View Full Product Details