Gene Transcripts

Gene Symbol
SIPA1L3
Entrez Gene ID
23094
Gene Name
signal induced proliferation associated 1 like 3
Gene Aliases
CTRCT45, SPAL3, SPAR3
Chromosome Location
Chr. 19: 37906823 - 38208372 on Build GRCh38
UniGene ID
Hs.655502
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015073.2 NP_055888.1 3 877
XM_005258671.4 XP_005258728.1 3 1318
XM_011526657.2 XP_011524959.1 3 1672
XM_017026517.1 XP_016882006.1 4 1772
XM_017026518.1 XP_016882007.1 3 1274
GenBank mRNA AY168880.1 - 1 623
BC036863.1 - 3 849
BC150620.1 - 3 856 9056990

View Full Product Details