Gene Transcripts

Gene Symbol
MRO
Entrez Gene ID
83876
Gene Name
maestro
Gene Aliases
B29, C18orf3
Chromosome Location
Chr. 18: 50795120 - 50825413 on Build GRCh38
UniGene ID
Hs.30495
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127174.1 NP_001120646.1 3, 4 327
NM_001127175.1 NP_001120647.1 3, 4 438
NM_001127176.1 NP_001120648.1 3, 4 438
NM_031939.3 NP_114145.2 4, 5 541
XM_005258365.3 XP_005258422.1 3, 4 351
XM_011526226.2 XP_011524528.1 4, 5 478
XM_017026035.1 XP_016881524.1 3, 4 318
XM_017026036.1 XP_016881525.1 4, 5 478
XM_017026037.1 XP_016881526.1 3, 4 467
GenBank mRNA AB042647.1 - 4, 5 405
AK054702.1 - 4, 5 475
AK294757.1 - 3, 4 282
AK295666.1 - 3, 4 439
AL705398.1 - 4, 5 449
BC029860.1 - 4, 5 428 5171860
DA416200.1 - 3, 4 293
DB473056.1 - 2, 3 176
DC333169.1 - 3, 4 440
DC403805.1 - 3, 4 323

View Full Product Details