Gene Transcripts

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
Chromosome Location
Chr. 3: 14124940 - 14143680 on Build GRCh38
UniGene ID
Hs.517817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024334.2 NP_077310.1 10, 11 1144
XM_017007176.1 XP_016862665.1 11, 12 1266
GenBank mRNA AK027466.1 - 4, 5 374
AK027757.1 - 10, 11 1070
AK027827.1 - 5, 6 427
AK027877.1 - 10, 11 1008
AK075010.1 - 10, 11 995
AK225589.1 - 10, 11 1067
AL136916.1 - 10, 11 1038
AY358625.1 - 10, 11 907
BC003125.1 - 10, 11 1039 3503636
BC008054.2 - 8, 9 634
BC011719.2 - 10, 11 1039 4079766
KU178769.1 - 9, 10 890
KU178770.1 - 8, 9 1464

View Full Product Details