Gene Transcripts

Gene Symbol
TXNDC15
Entrez Gene ID
79770
Gene Name
thioredoxin domain containing 15
Gene Aliases
C5orf14, UNQ335
Chromosome Location
Chr. 5: 134873770 - 134901633 on Build GRCh38
UniGene ID
Hs.741304
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024715.3 NP_078991.3 3 1284
GenBank mRNA AK026278.1 - 4 687
AK056512.1 - 3 1319
AK092425.1 - 1 1349
AK225454.1 - 4 687
AK225462.1 - 4 687
AK299578.1 - 3 577
AK307529.1 - 2 195
AY358649.1 - 3 712
BC001615.1 - 3 771 3544923
BC032568.1 - 3 685 5225890
BX538131.1 - 3 611

View Full Product Details