Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102396.1 | NP_001095866.1 | 4 | 571 | |
NM_025073.2 | NP_079349.2 | 4 | 559 | ||
NR_049741.1 | 4 | 621 | |||
NR_049742.1 | 3 | 453 | |||
GenBank mRNA | AI221103.1 | - | 3 | 278 | |
AK024821.1 | - | 4 | 549 | ||
AK126316.1 | - | 4 | 569 | ||
AK225734.1 | - | 4 | 550 | ||
BC005934.1 | - | 4 | 529 | 4249034 | |
CR749219.1 | - | 4 | 562 | ||
CR936771.1 | - | 4 | 562 | ||
DB459255.1 | - | 4 | 389 |