Gene Transcripts

Gene Symbol
TLR10
Entrez Gene ID
81793
Gene Name
toll like receptor 10
Gene Aliases
CD290
Chromosome Location
Chr. 4: 38772238 - 38782990 on Build GRCh38
UniGene ID
Hs.120551
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017388.2 NP_001017388.1 2 992
NM_001195106.1 NP_001182035.1 3 1309
NM_001195107.1 NP_001182036.1 3 1181
NM_001195108.1 NP_001182037.1 3 1212
NM_030956.3 NP_112218.2 4 1498
XM_011513760.2 XP_011512062.1 2 886
XM_011513761.2 XP_011512063.1 5 1579
XM_011513762.2 XP_011512064.1 4 1259
GenBank mRNA AB445680.1 - 1 860
AF296673.1 - 3 1346
AK292026.1 - 2 978
AK313468.1 - 2 1129
AY358300.1 - 2 970
BC089406.1 - 2 1344 3066588
BC109111.2 - 1 1467 40035401
BC109112.2 - 2 1010 40035404
DQ011502.1 - 1 860
DQ011503.1 - 1 860
DQ011504.1 - 1 860
DQ011505.1 - 1 860
DQ011506.1 - 1 860
DQ011507.1 - 1 860
DQ011508.1 - 1 860
DQ011509.1 - 1 860
DQ011510.1 - 1 860
DQ011511.1 - 1 860
DQ011512.1 - 1 860
DQ011513.1 - 1 860
DQ011514.1 - 1 860
DQ011515.1 - 1 860
DQ011516.1 - 1 860

View Full Product Details