Gene Transcripts

Gene Symbol
ODF3L2
Entrez Gene ID
284451
Gene Name
outer dense fiber of sperm tails 3 like 2
Gene Aliases
C19orf19
Chromosome Location
Chr. 19: 463346 - 490287 on Build GRCh38
UniGene ID
Hs.104777
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182577.2 NP_872383.1 4 832
XM_005259545.4 XP_005259602.1 4 1005
XM_017026671.1 XP_016882160.1 3 402
GenBank mRNA AK097378.1 - 4 832
BC104468.1 - 3 621 40032428
BC104469.1 - 4 725 40032432

View Full Product Details