Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004184.3 | NP_004175.2 | 5 | 896 | |
NM_173701.1 | NP_776049.1 | 5 | 672 | ||
NM_213645.1 | NP_998810.1 | 4 | 500 | ||
NM_213646.1 | NP_998811.1 | 4 | 724 | ||
XM_005268044.3 | XP_005268101.1 | 5 | 624 | ||
XM_006720249.3 | XP_006720312.1 | 6 | 858 | ||
XM_011537133.2 | XP_011535435.1 | 7 | 870 | ||
XM_011537135.2 | XP_011535437.1 | 6 | 754 | ||
XM_011537136.2 | XP_011535438.1 | 5 | 590 | ||
XM_017021627.1 | XP_016877116.1 | 6 | 762 | ||
XM_017021628.1 | XP_016877117.1 | 6 | 893 | ||
XM_017021629.1 | XP_016877118.1 | 7 | 868 | ||
GenBank mRNA | AK025487.1 | - | 3 | 300 | |
AK056100.1 | - | 5 | 1134 | ||
AK291141.1 | - | 4 | 476 | ||
AK300255.1 | - | 5 | 779 | ||
BC017489.1 | - | 5 | 772 | 3542671 | |
BC095453.1 | - | 6 | 783 | 30418785 | |
BG717903.1 | - | 4 | 505 | ||
BM561448.1 | - | 519 | |||
BX248006.1 | - | 5 | 616 | ||
M61715.1 | - | 5 | 702 | ||
M77804.1 | - | 4 | 557 | ||
X59892.1 | - | 5 | 626 | ||
X62570.1 | - | 4 | 577 |